Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Hydroxide (Pellets/Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
1 kg
3 kg
10 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Specifications
Specifications
pH | 13.5 |
Color | Yellow |
Boiling Point | 1320°C |
Melting Point | 360°C |
Assay Percent Range | ≥85.0% |
UN Number | 1813 |
Density | 2.04g/cm³ |
Quantity | 3 kg |
Formula Weight | 56.11 |
Percent Purity | ≥85.0% |
Show More |
Safety and Handling

Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Causes severe burns by all exposure routes. Water reactive. Use personal protective equipment. Do not allow contact with water. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. NFPA Health:3 Flammability:0 Instability:1 |
DOTInformation | DOT Class 8, : Corrosive |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction