Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide (Pellets/Certified ACS), Fisher Chemical™

Catalog No. P250500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 kg
3 kg
10 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P250-500 500 g Poly Bottle
P250-1 1 kg Poly Bottle
P250-3 3 kg Poly Bottle
P250-10 10 kg Poly Pail
P250-50 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. P250-500
Only null left
Add to Cart

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

pH 13.5
Color Yellow
Boiling Point 1320°C
Melting Point 360°C
Assay Percent Range ≥85.0%
UN Number 1813
Density 2.04g/cm³
Quantity 500 g
Formula Weight 56.11
Percent Purity ≥85.0%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Physical Form Solid
Assay ≥85.0%
Chemical Name or Material Potassium Hydroxide
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes severe burns by all exposure routes. Water reactive. Use personal protective equipment. Do not allow contact with water. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:1
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.