Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentane (HPLC), Fisher Chemical™
GSA_VA

Catalog No. P3994 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass
Safe-Cote™ Amber Glass 
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P399-4 4 L Amber Glass
P399-1 1 L Amber Glass
P399SK-1 1 L Safe-Cote™ Amber Glass 
P399SK-4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. P399-4
  • $582.00
    $291.65 / Each

    Save $290.35

    50% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,789.00
    $896.65 / Case of 4 EA

    Save $892.35

    50% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
 Restricted Item
Add to Cart

missing translation for 'validMainframeMsg'

Submicron filtered

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Spécifications

Melting Point -130°C
Absorbance 0.014 AU max. at 254nm, 0.1 AU max. at 210nm, 0.3 AU max. at 200nm, 1.0 AU max. at 190nm
Density 0.626g/cm³
Quantity 4 L
Identification Pass Test
Packaging Amber Glass
Physical Form Liquid
Refractive Index 1.3540 to 1.3570 (at 25°C)
Residue after Evaporation 5ppm max.
Assay Percent Range >99%
Vapor Pressure 573mbar at 20°C
Linear Formula CH3(CH2)3CH3
Viscosity 0.25 mPa/s at 20°C
UN Number 1265
Water 0.02% max.
Boiling Point 36°C
Formula Weight 72.15
Percent Purity >99%
Grade Mobile phase for HPLC applications
Chemical Name or Material Pentane
Afficher plus de résultats Afficher moins de résultats
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause skin, eye, and respiratory tract irritation. Aspiration hazard if swallowed - can enter lungs and cause damage. Repeated exposure may cause skin dryness or cracking. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:1
Flammability:4
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.