Learn More
Sodium Sulfate Anhydrous, Certified ACS, Suitable for Pesticide Residue Analysis (Granular, 10-60 mesh), Fisher Chemical™
Shop All Fisherbrand ProductsSpecifications
Specifications
| Melting Point | 884°C |
| Color | White |
| pH | 5 to 8 |
| Physical Form | Solid |
| Quantity | 1 kg |
| Assay Percent Range | ≥99.0 % |
| Formula Weight | 142.04 |
| Percent Purity | ≥99.0% |
| Grade | Suitable for pesticide residue analysis |
| Identification | Pass Test |
| Show More |
Safety and Handling
| Hazard Category | SIGNAL WORD: CAUTION! |
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic. NFPA Health: 1 Flammability: 1 Instability: 0 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.