Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid (Gerber), Fisher Chemical

Catalog No. SA1764 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
SA176-4 4 L Poly Bottle
1 options

Catalog No. SA176-4

Only null left
Add to Cart

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0.3
Color Colorless to Brown
Melting Point 10°C
Boiling Point 290°C
Density 1.84g/cm³
Identification Pass Test
Packaging Poly Bottle
Specific Gravity 1.82 to 1.825 (at 25°C)
Vapor Pressure <0.001 mmHg at 20°C
Assay Percent Range 91.0 to 92.25 %
Quantity 4 L
Physical Form Liquid
UN Number 1830
Percent Purity 91.0 to 92.25%
Chemical Name or Material Sulfuric Acid, Gerber
Show More Show Less
DOTInformation Class 8: Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.