Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide Solution, 45% w/w (Certified), Fisher Chemical™

Catalog No. SP236200 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Poly Bottle
Poly Drum
Poly Pail
PolyPac™
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP236200 200 L Poly Drum
SP236-500 500 mL Poly Bottle
SP236-4 4 L Poly Bottle
SP236-20 20 L Poly Pail
SP236P-20 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. SP236200
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

pH 12.0
Color Colorless
Melting Point -28.8°C
Assay Percent Range 45 to 47 %
Density 1.510g/cm³
Quantity 200 L
Formula Weight 56.11
Percent Purity 45 to 47%
Grade Certified
Concentration or Composition (by Analyte or Components) 45%
Identification Pass Test
Packaging Poly Drum
Vapor Pressure 2.6mmHg at 20°C
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide Solution, 45% w/w
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.