Learn More
Thermo Scientific™ KingFisher™ Plastics for 24 deep-well format
Description
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
| Product Type | 6-tip comb and 24 DW plate |
| Format | Plate and Comb |
| For Use With (Equipment) | KingFisher Duo Prime |
| Quantity | 48 Pc. |
Frequently Asked Questions (FAQs)
Do not use the tip combs if the clips are bent or missing. Not having clips can cause the tip comb to drop from the holder during the run, which would cause the run to abort. If you received tip combs with bent or broken retaining clips, you can contact Technical Support at techsupport@thermofisher.com to request a free-of-charge replacement.
The KingFisher Duo Purification System consumables are listed on the following website and they vary depending on whether you are using the 6- or 12-pin magnet: https://www.thermofisher.com/order/catalog/product/97003500
The KingFisher Duo Prime Purification System has the following additional features compared to the KingFisher Duo Purification System:
• Improved safety shield design
• UV lamp
• Handheld barcode reader option
• Additional language options
• Optimized elution strip block for better alignment of the plastic strip
The KingFisher Duo can run any script written for the KingFisher Duo Prime and does not require additional modifications.
For research use only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.