Learn More
Thermo Scientific™ KingFisher™ Plastics for 24 deep-well format
Catalog No. 14387357
Change view
Click to view available options
Product Type:
24 Combi tip comb
24 DW plate, barcoded, sterile
24 DW tip comb and plate
24 DW tip comb, plate, barcode
24 DW tip comb, plate, sterile
24 Deep Well plate, barcoded
24 Deep-well Plate
24 Deep-well Plate, Sterile
6-tip comb and 24 DW plate
For Use With (Equipment):
KF Duo Prime, Flex, Presto
KingFisher Apex
KingFisher Duo Prime
KingFisher Flex and Presto
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
14-387-357 24 DW tip comb, plate, sterile KingFisher Apex
97-002-580 24 Combi tip comb KingFisher Apex
22-387-034 24 Deep-well Plate KF Duo Prime, Flex, Presto
22-387-035 24 Deep-well Plate, Sterile KF Duo Prime, Flex, Presto
22-387-033 24 DW tip comb and plate KingFisher Flex and Presto
14-387-739 24 DW tip comb, plate, sterile KingFisher Flex and Presto
14-387-292 6-tip comb and 24 DW plate KingFisher Duo Prime
14-387-351 24 Deep Well plate, barcoded KingFisher Apex
14-387-356 24 DW tip comb, plate, barcode KingFisher Apex
14-387-352 24 DW plate, barcoded, sterile KingFisher Apex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 14-387-357 Supplier Thermo Scientific™ Supplier No. 97002620B
Add to Cart
Add to Cart
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Product Type 24 DW tip comb, plate, sterile
For Use With (Equipment) KingFisher Apex
Quantity 50/Cs.
I received the KingFisher Flex/KingFisher Duo Prime tip combs and the retaining clips are bent or broken off. When I try to unbend the retaining clips, they break. Can I still use the tip combs if the retaining clips are bent or broken off?
Do not use the tip combs if the clips are bent or missing. Not having clips can cause the tip comb to drop from the holder during the run, which would cause the run to abort. If you received tip combs with bent or broken retaining clips, you can contact Technical Support at techsupport@thermofisher.com to request a free-of-charge replacement.
For research use only. Not for use in diagnostic procedures.