Learn More
Thermo Scientific™ KingFisher Apex Accessories
Description
KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality
- User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
- Provides versatile and high-speed purification with reproducible results
- Easy to use interface with touch screen empowering the user to control all details of the application and purification process
- BindIx PC Software for added convenience
- Extensive selection of validated protocols for different type of applications
- Cloud library for quick protocol retrieval
Specifications
Specifications
| Product Type | 24 DW Heating Block |
| Format | 24 Deep-well Heating Block |
| Capacity | 24 Samples |
| Disposable | Reusable |
| For Use With (Equipment) | KingFisher Apex |
| Quantity | 1 Pc. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.