Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ KingFisher Apex Accessories
GSA_VA

Catalog No. 24079930
Encompass
Encompass_Preferred
Change view
Click to view available options
Product Type:
24 Combi Head
24 DW Heating Block
96 DW Head
96 DW Heating Block
96 KF Heating Block
96 PCR Head
Combi Head
Heating Block 24
Heating Block 96
Matrix Heating Block
PCR Plate Heating Block
Plate Adapter
Storage Tube Adapter
UV Lamp (Replacement)
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
24-079-930 96 DW Head
24-075-970 Matrix Heating Block
24-079-910 96 PCR Head
24-079-920 Combi Head
24-079-940 24 Combi Head
24-075-910 PCR Plate Heating Block
24-075-920 96 KF Heating Block
24-075-930 96 DW Heating Block
24-075-940 24 DW Heating Block
24-075-950 Heating Block 96
24-075-960 Heating Block 24
N21445 Storage Tube Adapter
N21446 Plate Adapter
N21447 UV Lamp (Replacement)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 24-079-930 Supplier Thermo Scientific™ Supplier No. 24079930
Add to Cart
Edge
Add to Cart
Request a Quote

KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source.

KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality

  • User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
  • Provides versatile and high-speed purification with reproducible results
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • BindIx PC Software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type 96 DW Head
Volume (Metric) Processing 96 KF plate 50 to 150 μL, 96 DW plate 50 to 1000 μL
Format 96 Deep-well Head
Capacity 96 Samples
For Use With (Equipment) KingFisher Apex
Magnetic Rods 96
Particle Size (Metric) ca. >1 μm
Type of Magnet Head 96 DW
Plate Type 96 KF, 96 DW
How many kinds (type or size) of magnets are there for the KingFisher Apex 96 DW Head (Cat. No. 24079930)?

Please see the Thermo Scientific KingFisher Flex User Manual, page 32. As you will see, there are four different heads. In terms of magnet force, it's a permanent magnet so the expected life is >100 years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.