Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KG 5
GREENER_CHOICESDP

Catalog No. 526050
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-605-0 50 mg
52-601-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-605-0 Supplier Tocris Bioscience™ Supplier No. 5260/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PDGFRβ, B-Raf, c-Raf, FLT3 and KIT inhibitor

KG 5 is a PDGFRβ and B-Raf allosteric inhibitor. Also inhibits c-Raf, FLT3 and KIT. Disrupts blood vessel formation in zebrafish embryos and inhibits angiogenesis in a mouse angiogenesis model. Inhibits growth of renal and pancreatic tumors in mice. Also arrests cells in prometaphase. Orally available.

Specifications

Chemical Name or Material 2-(Methylthio)-6-[4-[5-[[3-(trifluoromethyl)phenyl]amino]-1H-1,2,4-triazol-3-yl]phenoxy]-4-pyrimidinamine
CAS 877874-85-6
Quantity 50 mg
Target Raf Kinase Inhibitors
Molecular Formula C20H16F3N7OS
Formula Weight 459.45
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.