Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KGA10 Hooded Coveralls, Elastic Wrists and Ankles
GREENER_CHOICE

Catalog No. 17101151 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-101-151 Medium
17-101-152 Large
17-101-153 X-Large
17-101-154 2X-Large
17-101-155 3X-Large
17-101-156 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-101-151 Supplier Ansell Supplier No. 67308
Add to Cart
Add to Cart
This item is not returnable. View return policy

Light-duty coveralls offer protection against dirt and grime. Popular for agriculture, food processing, maintenance, labs, and dusty environments.

  • For non-hazardous particulate protection
  • Lightweight and highly breathable spunbond fabric allows moisture to escape for maximum comfort
  • Hooded, elastic wrists and ankles
  • Not made with natural rubber latex or silicone
  • Recyclable with the RightCycle™ Program
TRUSTED_SUSTAINABILITY

Specifications

Product Type Light-duty Coveralls
Size Medium
Gender Unisex
Material Spunbond Polypropylene
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Front Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Dust
Certifications/Compliance ANSI/ISEA 101-2014
Certifications/Compliance (Europe) CE Category I PPE
Latex Free Yes
Model KleenGuard™ KGA10
For Use With (Application) Agriculture, Food Processing, General Maintenance, Laboratories
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.