Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KGA20 Hooded Coveralls
GREENER_CHOICE

Catalog No. 17101172 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-101-172 4X-Large
17-101-167 Medium
17-101-168 Large
17-101-169 X-Large
17-101-170 2X-Large
17-101-171 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-101-172 Supplier Ansell Supplier No. 68976
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Coveralls with attached hood provide lightweight protection against heavy dust and limited splashes.

  • Comfortable, lightweight, SMS fabric is breathable and allows moisture to escape
  • Hooded, Elastic Wrist and Ankle Design
  • Not made with natural rubber latex or silicone
  • Recyclable with the RightCycle™ Program
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 4X-Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Limited-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Dust, Liquid
Certifications/Compliance ANSI/ISEA 101-1996
Certifications/Compliance (Europe) CE Category I PPE
Latex Free Yes
Model KleenGuard™ KGA20
For Use With (Application) Agriculture, Food Processing, General Maintenances, Laboratories
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.