Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KGA20 Hooded Coveralls with Boots
GREENER_CHOICE

Catalog No. 17101173 Shop All Ansell Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-101-173 Large
17-101-174 X-Large
17-101-175 2X-Large
17-101-176 3X-Large
17-101-177 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-101-173 Supplier Ansell Supplier No. 68977
Add to Cart
Add to Cart
This item is not returnable. View return policy

Coveralls with attached hood and boots provide lightweight protection against heavy dust and limited splashes.

  • For Non-Hazardous Particulate Protection
  • Comfortable, lightweight, SMS fabric is breathable and allows moisture to escape
  • Hood and Boot, Elastic Wrist and Ankle Design
  • Not made with natural rubber latex or silicone
  • Recyclable with the RightCycle™ Program
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Limited-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper
Sterility Non-sterile
Resistance Dust, Liquid
Certifications/Compliance ANSI/ISEA 101-1996
Certifications/Compliance (Europe) CE Category I PPE
Latex Free Yes
Model KleenGuard™ KGA20
For Use With (Application) Agriculture, Food Processing, General Maintenances, Laboratories
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.