Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Khellin 95.0+%, TCI America™
SDP

Catalog No. K003925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K003925G 25 g
K00395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K003925G Supplier TCI America Supplier No. K003925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82-02-0
Molecular Formula C14H12O5
Molecular Weight (g/mol) 260.245
MDL Number MFCD00005007
InChI Key HSMPDPBYAYSOBC-UHFFFAOYSA-N
Synonym khellin, visammin, amicardine, methafrone, coronin, amikellin, ammipuran, ammivisnagen, benecardin, corafurone
PubChem CID 3828
ChEBI CHEBI:6133
IUPAC Name 4,9-dimethoxy-7-methylfuro[3,2-g]chromen-5-one
SMILES CC1=CC(=O)C2=C(C3=C(C(=C2O1)OC)OC=C3)OC

Specifications

Melting Point 152°C
Color Yellow
Quantity 25 g
UN Number 2811
Formula Weight 260.25
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Khellin
RTECSNumber LV1050000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.