Learn More
Medchemexpress LLC HY-101140A 1mg Medchemexpress, KI696 isomer CAS:1799974-69-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101140A1MG
Encompass_Preferred
Medchemexpress, HY-101140A 1mg KI696 isomer CAS:1799974-69-8 KI696 isomer is the less active isomer of KI696. KI696 is a high affinity probe that disrupts the Keap1/NRF2 interaction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2050
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.