Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA0082 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7140108
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-7342 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-7342 Supplier Bethyl Laboratories, Inc Supplier No. A300304AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to

FTSJD2 is a S-adenosyl-L-methionine-dependent methyltransferase that mediates mRNA cap1 2′-O-ribose methylation to the 5′-cap structure of mRNAs. It methylates the ribose of the first nucleotide of a m7GpppG-capped mRNA to produce m7GpppNmp (cap1). Cap1 modification is linked to higher levels of translation. It may be involved in the interferon response pathway.

Specifications

Antigen KIAA0082
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CMTR1
Gene Accession No. Q8N1G2, Q9DBC3
Gene Alias FtsJ methyltransferase domain containing 2, KIAA0082, MTr1, hMTr1, FTSJD2
Gene Symbols CMTR1
Host Species Rabbit
Immunogen Between 100 and 150
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23070, 74157
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.