Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA0087 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734648 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4648 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4648 Supplier Proteintech Group Inc Supplier No. 249731AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This KIAA0087 antibody gives a 40 kDa band in Western Blot.

KIAA0087 (KIAA0087 LncRNA) is an RNA Gene, and is affiliated with the non-coding RNA class. Diseases associated with KIAA0087 include Endometrial Cancer and Pancreatic Cancer.

Specifications

Antigen KIAA0087
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene KIAA0087
Gene Accession No. Q14695
Gene Alias KIAA0087
Gene Symbols KIAA0087
Host Species Rabbit
Immunogen KIAA0087 Fusion Protein Ag21759
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9808
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.