Learn More
Invitrogen™ KIAA1529 Polyclonal Antibody

Description
KIAA1529 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | KIAA1529 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CCDC180 |
| Gene Accession No. | Q9P1Z9 |
| Gene Alias | BDAG1; Behcet's Disease Associated Gene 1; C9orf174; CCDC180; coiled-coil domain containing 180; coiled-coil domain-containing protein 180; E230008N13Rik; Gm1025; KIAA1529; RIKEN cDNA E230008N13 gene |
| Gene Symbols | CCDC180 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.