Learn More
Sigma Aldrich Fine Chemicals Biosciences HIS SelectR Nickel Magneti
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H99145ML
Encompass
HIS-Select Magnetic Agarose Beads consist of paramagnetic immobilized metal-ion affinity chromatography (IMAC) resin that contain a proprietary quadridentate chelate which is bound with nickel and covalently attached through a non-charged hydrophilic linker to magnetic beaded agarose. The magnetic properties of the beads aid in manipulations such as repetitive washings and recovery of the protein bound beads. This leads to greater experimental reproducibility and more accurate quantitation of the His-tagged proteins of interest.
Catalog No. 50-291-4258
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.