Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Celestron™ Kids Microscope Kits
SCI_ED

Catalog No. S81332 Shop All Celestron International Products
Change view
Click to view available options
Model:
28 Piece Kit
Basic Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
S81332 Basic Kit
S81331 28 Piece Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning: Contains Chemicals Listed as Cancer and Reproductive Harm in the State of California
Catalog No. S81332 Supplier Celestron™ Supplier No. 44124
Warning: Contains Chemicals Listed as Cancer and Reproductive Harm in the State of California

Celestron™ Kids Microscope Kits include all of the essential items needed to begin exploring the wonders of the microscopic world

Basic Kit

  • Up to 1200x power
  • Microscope kit includes 100x, 600x, and 1200x objectives
  • Bonus 2x magnifying glass included
  • Additional accessories includes 7 blank slides with covers
  • CPSIA and ASTM F963 tested and compliant
  • Age graded 8+

28 Piece Kit

  • Beginner set with metal bodied biological microscope
  • 3 objective lenses for high power viewing - 100x, 600x, and 1200x
  • Form fitted carry case for portability and protection
  • 7 blank slides, 5 prepared specimen slides, and 7 slide cover slips and labels
  • Accessories to get you experimenting right away: plastic scalpel, tweezers, probe, 2x magnifier, and specimen samples
  • CPSIA and ASTM F963 tested and compliant
  • Age graded 8+

Specifications

Product Type Microscope
Model Basic Kit
Lighting Type Adjustable Illuminator
Includes 7 Reusable Blank Slides/Covers, 2x Hand Lens
Magnification Power 1200X
Objective Types Achromatic
For Use With (Application) Ages 8+
Warranty 2 Years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.