Learn More
Celestron™ Kids Pocket Zoom Microscope
SCI_ED
Catalog No. S81336 Shop All Celestron International Products
Change view
Click to view available options
Magnification Power:
60X to 120X
Catalog No. Magnification Power
S81336 60X to 120X
1 options
Warning: Contains Chemicals Listed as Cancer and Reproductive Harm in the State of California
Catalog No. S81336 Supplier Celestron™ Supplier No. 44116
Add to Cart
Add to Cart
Warning: Contains Chemicals Listed as Cancer and Reproductive Harm in the State of California
Celestron™ Kids Pocket Zoom Microscope is pocket sized and can be carried anywhere!
  • Great for viewing 3D objects and prepared slides
  • LED illuminator adds more light when needed
  • Zoom from 60x to 120x
  • CPSIA and ASTM F963 tested
  • Age rated 8+
Specifications
Eyepiece Fixed
Head Style Monocular
Lighting Type LED, Bright White
Description Requires AAA Batteries (1)
Product Type Microscope
Magnification Power 60X to 120X
Objective Types Fixed
For Use With (Application) Ages 8+
Weight (English) 5.5 oz.
Weight (Metric) 181 g
Dimensions (L x W x H) 2.5 x 0.9 x 0.9 in.