Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kieselguhr, Purified, Calcined, MilliporeSigma™

Catalog No. M1079101000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
1 kg
25 kg
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1079101000 1 kg Plastic Bottle
M1079100250 250 g Plastic Bottle
M1079109025 25 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1079101000 Supplier MilliporeSigma Supplier No. 1.07910.1000
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]

Specifications

Color White
Physical Form Solid
Quantity 1 kg
Solubility Information <0.001g/L
Packaging Plastic Bottle
EINECSNumber 272-489-0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.