Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF15 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501563272
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3272 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3272 Supplier Bethyl Laboratories, Inc Supplier No. A302707AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Plus-end directed kinesin-like motor enzyme involved in mitotic spindle assembly.

Specifications

Antigen KIF15
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene KIF15
Gene Accession No. Q9NS87
Gene Alias HKLP2, kinesin-like 7, kinesin-like protein 2, kinesin-like protein 7, KNSL7, NY-BR-62, serologically defined breast cancer antigen NY-BR-62
Gene Symbols KIF15
Host Species Rabbit
Immunogen Between 1338 and 1388
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56992
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.