Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF20A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120355
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120355 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120355 Supplier Invitrogen™ Supplier No. PA5120355
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, A-549, Mouse spleen, Mouse testis, Mouse heart, Mouse lung, Mouse brain, Rat spleen. The target is usually found in the following locations: Cytoplasm, Golgi apparatus, cytoskeleton, spindle. Immunogen sequence: EICNEMVEQM QQREQWCSEH LDTQKELLEE MYEEKLNILK ESLTSFYQEE IQERDEKIEE LEALLQEARQ QSVAHQQSGS ELALRRSQRL AASASTQQLQ EVKAKLQQCK AELNSTTEEL HKYQKMLEPP PSAKPFTIDV DKKLEEGQKN IRLLRTELQK LGESLQSAER ACCHSTGAGK LRQALTTCDD ILIKQDQTLA ELQNNMVLVK LDLRKKAACI AEQYHTVLKL QGQVSAKKRL GTNQENQQPN QQPPGKKPFL RNLLPRTPTC QSSTDCSPYA RILRSRRSPL LKSGPFGKKY.

Mitotic kinesin required for chromosome passenger complex (CPC)-mediated cytokinesis. Following phosphorylation by PLK1, involved in recruitment of PLK1 to the central spindle. Interacts with guanosine triphosphate (GTP)-bound forms of RAB6A and RAB6B. May act as a motor required for the retrograde RAB6 regulated transport of Golgi membranes and associated vesicles along microtubules. Has a microtubule plus end-directed motility. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF20A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene KIF20A
Gene Accession No. O95235, P97329
Gene Alias AA415432; GG10_2; KIF20A; kinesin family member 20A; kinesin-like protein 174; Kinesin-like protein KIF20A; mitotic kinesin-like protein 2; MKlp2; RAB6 interacting, kinesin-like (rabkinesin6); rab6-interacting kinesin-like protein; RAB6KIFL; Rabkinesin-6
Gene Symbols KIF20A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 591-890 of human KIF20A (NP_005724.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10112, 19348, 361308
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.