Learn More
KIF20A/RAB6KIFL Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300879AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
Mitotic kinesin required for chromosome passenger complex (CPC)-mediated cytokinesis. Following phosphorylation by PLK1, involved in recruitment of PLK1 to the central spindle. Interacts with guanosine triphosphate (GTP)-bound forms of RAB6A and RAB6B. May act as a motor required for the retrograde RAB6 regulated transport of Golgi membranes and associated vesicles along microtubules. Has a microtubule plus end-directed motility.Specifications
| KIF20A/RAB6KIFL | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| O95235 | |
| KIF20A | |
| Between 840 and C-term | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| KIF20A | |
| MKLP2, RAB6KIFL, GG10_2, RAB6 interacting, kinesin-like (rabkinesin6), kinesin-like protein KIF20A, mitotic kinesin-like protein 2, rab6-interacting kinesin-like protein, rabkinesin-6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10112 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.