Learn More
KIF20A/RAB6KIFL Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
Specifications
Specifications
| Antigen | KIF20A/RAB6KIFL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | KIF20A |
| Gene Accession No. | O95235 |
| Gene Alias | MKLP2, RAB6KIFL, GG10_2, RAB6 interacting, kinesin-like (rabkinesin6), kinesin-like protein KIF20A, mitotic kinesin-like protein 2, rab6-interacting kinesin-like protein, rabkinesin-6 |
| Gene Symbols | KIF20A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.