Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIF26B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555901
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555901 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555901 Supplier Invitrogen™ Supplier No. PA555901
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GYIPMKTNIT VYPCIAMSPR NIQEPEAPTA TPKAGPTLAQ SRESKENSAK KEMKFEDPWL KREEEVKKET AHPNEEGMMR CETATGPSNA ETR Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 65%.

KIF26B is a member of kinesin-11 family and plays important role in embryogenesis. It is a downstream target of Sall1 and is essential for kidney development. KIF26B is also involved in tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF26B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Kif26b
Gene Accession No. Q2KJY2
Gene Alias 4832420M10; BC056349; D230039L06Rik; Kif26b; kinesin family member 26B; kinesin family protein 26b; kinesin-like protein KIF26B; N-11 kinesin; RGD1560022; RGD1560572
Gene Symbols Kif26b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIF26B. Recombinant protein control fragment (Product #RP-94416).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55083
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.