Learn More
KIFC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
This antibody was raised against the C-terminal region of human KIFC1 protein, and recognizes 55-70 kDa of endogenous KIFC1 protein in multiple tissue lysates.
Specifications
Specifications
| Antigen | KIFC1 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | KIFC1 |
| Gene Accession No. | Q5XI63, Q9BW19, Q9QWT9 |
| Gene Alias | HSET, KIFC1, kinesin family member C1, Kinesin like protein 2, Kinesin like protein KIFC1, Kinesin related protein HSET, KNSL2 |
| Gene Symbols | KIFC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.