Learn More
DWK Life Sciences Kimble™ KIMAX™ Heavy-Duty Graduated Centrifuge Tubes with Screw Caps
SDP
Catalog No. 03341661 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10 mL
40 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
03-341-661 10 mL
03-341-662 40 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-341-661 Supplier DWK Life Sciences Supplier No. 4520010
Add to Cart
Add to Cart
This Standard Taper KIMAX™ tube is a heavy-duty version of 45166.
  • Graduated and calibrated to contain
  • Scale is durable white ceramic enamel
  • Made with heavier walled tubing to withstand higher centrifugation speed
  • Top has a screw thread finish
  • Autoclavable screw-thread cap with PTFE-faced rubber liner is included but not attached
  • Replacement cap is 45066C
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 10 mL
Product Type Centrifuge Tube with Screw Cap