Learn More
Ansell A8 Ankle-High Shoe Covers with Traction Strips
GREENER_CHOICE
Catalog No. 19065703 Shop All Ansell Products
Change view
Click to view available options
Size:
Universal
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-065-703 X-Large
19-188-584 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-065-703 Supplier Ansell Supplier No. 69353
Add to Cart
Add to Cart
This item is not returnable. View return policy
Shoe covers fit over the ankle with elastic and extend to mid-calf for extra protection from moderate fluid contact. Adhesive traction strips on the bottom help prevent slips.
  • Three layers of SMS fabric
  • Adhesive traction strips
  • For moderate fluid contamination
  • Latex free
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Material SMS
Latex Free Yes
Color Blue
Disposable Single-use
Sterility Non-sterile
Sole Type Traction Strips
Model 69353
Thickness 3-ply
Includes Adhesive traction stripes
Resistance Fluid
Product Type Shoe Covers