Learn More
Ansell Kimtech™ A8 Certified Lab Coats with Knit Cuffs and Collar
GREENER_CHOICE
Catalog No. 19022847 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Color:
Blue
White
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-022847 Large Blue
18-022-840 Small White
18-022-841 Medium White
18-022-842 Large White
18-022-843 X-Large White
19-022844 2X-Large White
19-022850 2X-Large Blue
18-022-849 X-Large Blue
19-022846 Medium Blue
19-022845 Small Blue
19-188-595 3X-Large Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 19-022847 Supplier Ansell Supplier No. 10032
Add to Cart
Edge
Add to Cart
Breathable SMS lab coats protect from light fluid contact and feature soft knit cuffs and 3 pockets. Choose from white or blue colors.
  • Lab coats with knit cuffs and collar
  • Comfort and protection from light fluid contact
  • 3-layer SMS nonwoven fabric
  • Soft knit cuffs and collar
  • Mid-calf length
  • Three reinforced pockets and five snaps
  • Neck loop for convenient hanging
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material SMS
Color Blue
Closure Type Snaps
Cuff Style Knit
Disposable Limited-use
Sterility Non-sterile
Style Mid-calf Length
No. of Pockets 3
Collar Type Knit
Model 10032
Size (English) Chest 42 to 46 in.
Thickness 3-ply
Includes Hanging Neck Loop
Construction 3-ply SMS
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.