Learn More
Kimberly-Clark™ Professional Jackson Safety™ BC100 Bump Caps
Description
- Lightweight for comfort
- Made of molded HDPE (high-density polyethylene)
- 4-point injection molded system includes an absorbent brow pad
- Pinlock sizing system can accommodate head sizes 6.5 to 8.25
- Can be made compatible with a face shield for additional coverage
- Replacement sweat bands and suspension inserts available
Specifications
Specifications
| Color | Blue |
| Material | HDPE |
| Product Type | Low-profile Bump Cap |
| Quantity | 12/Pk. |
| Size | Adjustable (Pin-lock sizing) |
| Suspension Type | 4-Point Injection Molded |