Learn More
Kimberly-Clark™ Professional Jackson Safety™ BC100 Bump Caps
Kimberly-Clark™ Professional Jackson Safety™ BC100 Bump Caps offer protection from minor bumps and scalp lacerations
Supplier: Kimberly-Clark™ Professional 14812
Description
- Lightweight for comfort
- Made of molded HDPE (high-density polyethylene)
- 4-point injection molded system includes an absorbent brow pad
- Pinlock sizing system can accommodate head sizes 6.5 to 8.25
- Can be made compatible with a face shield for additional coverage
- Replacement sweat bands and suspension inserts available
Specifications
Green | |
Low-profile Bump Cap | |
Adjustable (Pin-lock sizing) |
HDPE | |
12/Pk. | |
4-Point Injection Molded |