Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ G10 Flex Nitrile Gloves

Catalog No. 19005429 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-005-429 X-Large
19-005-425 X-Small
19-005-426 Small
19-005-427 Medium
19-005-428 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-005-429 Supplier Ansell Supplier No. 38522
This item is not returnable. View return policy

Thin mil gloves protect without sacrificing productivity. Suitable for food handling and general purpose applications.

  • Powder free, nitrile
  • Textured fingertips give workers dexterity and control for delicate tasks
  • Touch screen compatible
  • Ambidextrous
  • Beaded cuff
  • Ideal for food handling, precision electronic assembly, painting, automotive and small parts handling
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance FDA 21 CFR 170-199
Applications Industrial
AQL 1.5
Color Blue
Exterior Finish Partially Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Large
Thickness (English) 2 mil
Thickness (Metric) 0.05 mm
Model 38522
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.