Learn More
Ansell KleenGuard™ G40 Smooth Nitrile Coated Multi-Purpose Gloves
Catalog No. 19005424 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-005-424 2X-Large
19-005-420 Small
19-005-421 Medium
19-005-422 Large
19-005-423 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-005-424 Supplier Ansell Supplier No. 38432
This item is not returnable. View return policy
General purpose gloves designed for conditions which require protection, strength, grip and dexterity in industrial environments. Nitrile coating offers a secure grip while the seamless knit nylon back offers a comfortable fit.
  • Designed for conditions which require protection, strength, grip and dexterity in industrial environments
  • Nitrile coating enhances grip capabilities and protects against liquid permeation
  • Seamless knit nylon back is comfortable and breathable
  • Dark color helps to conceal dirt for extended wear
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Nitrile
Color Black
Exterior Finish No Texture
Material Nylon
Product Type Seamless Knit Gloves
Size 2X-Large
Glove Cuff Style Knit
Series Kleengaurd™ G40
Length (English) 10 in.
Length (Metric) 25.4 cm