Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ G60 Knuckle Coated High-Visibility Cut-Resistant Gloves

Catalog No. 19005419 Shop All Ansell Products
Change view
Click to view available options
Length (English):
8.4 in.
8.9 in.
9.6 in.
10 in.
Length (Metric):
21.3 cm
22.6 cm
24.3 cm
25.4 cm
Size:
Small
Medium
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Length (Metric) Size
19-005-419 10 in. 25.4 cm 2X-Large
19-005-415 8.4 in. 21.3 cm Small
19-005-416 8.9 in. 22.6 cm Medium
19-005-418 9.6 in. 24.3 cm X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-005-419 Supplier Ansell Supplier No. 38651
This item is not returnable. View return policy

High-visibility cut-resistant gloves with polyurethane coating are ideal for working in environments that pose a risk of minor cuts and abrasions.

  • EN 388 Level 3 / ANSI Level 2 Cut Resistance
  • EN 388 Level 4 abrasion and Level 4 tear resistance
  • Certified to meet the EN 420 standards
  • Bright orange fiber aids in overall safety compliance and awareness of how close a hand is in proximity to potential dangers
  • Made of ultra-high molecular weight polyethylene (UHMWPE) yarn with nylon and stretch fiber, which provides both cut resistance and a comfortable fit
  • Polyurethane coating enhances dry and oil grip and hides dirt for longer wear
  • Dark coating helps hide dirt for longer wear
  • Each pair has a dedicated right and left glove for a snug fit
  • Knuckle coating helps keep hands and gloves protected from liquids and oils to enforce compliance and improve glove longevity
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420
Color Black, Orange
Cut Resistance Rating ANSI A3
Material Dyneema, Nylon, Lycra
Product Type Cut-resistant Gloves
Size 2X-Large
EN388 Standard EN 388
For Use With (Application) Metal fabrication, Glass handling, automotive assembly, stamping and assembly
Glove Cuff Style Knit
Model 38651
Series Kleenguard™ G60
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.