Learn More
Ansell KleenGuard™ Nemesis VL™ Safety Glasses
GREENER_CHOICE
Catalog No. 19706011 Shop All Ansell Products
Change view
Click to view available options
Lens Tint:
Clear
Indoor/Outdoor
Smoke Mirror
Lens Coating:
Anti-fog
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Coating
19-706-011 Indoor/Outdoor -
19-706-008 Clear -
19-706-009 Clear Anti-fog
19-706-010 Smoke Mirror -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-706-011 Supplier Ansell Supplier No. 29112
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Neck cord
Frameless ultra-lightweight design
  • No-brow design increases upward and peripheral vision
  • Slip-stop temples designed to prevent slipping and sliding when hot
  • Compact profile accommodates smaller faces
  • 99.9% UV protection
  • Base curve 8
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets ANSI Z87.1+ standards for high impact (29112-00 meets ANSI Z87.1+2010 and 29112-01 meets ANSI Z87.1+2015)
Lens Tint Indoor/Outdoor
Frame Color Gun Metal
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.