Description
- Durable, lightweight construction
- Offer a sport design that fits smaller, slimmer heads without adjustments
- Features a stronger nose piece and a wider bridge for enhanced comfort
- Polycarbonate lenses that provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
- Wraparound frame has a base curve of 8 that extends the range of peripheral protection
- Each pair comes with a neck cord for securing, so they are ready at a moment’s notice
- Manufactured to comply with the ANSI / ISEA Z87.1+ standard for personal eye protection
- Feature a sporty, flexible, lightweight design
- Patented FLEX-DRY designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose
Specifications
Specifications
| Lens Coating | Anti-fog |
| Certifications/Compliance | Meets ANSI Z87.1+ standards for high impact (38474-00 and 38474-01 meets ANSI Z87.1+2010 and 38474-02 meets ANSI Z87.1+2015) |
| Lens Tint | Amber |
| Product Type | Safety Eyewear |
| Frame Color | Black |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | Nylon |
| UV Protection | 99.9% UVA/UVB/UVC |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.