Learn More
KleenGuard™ V50 Calico™ Safety Eyewear
Catalog No. 19170100 Shop All Ansell Products
Change view
Click to view available options
UV Protection:
UV400
Catalog No. UV Protection
19-170-100 UV400
1 options
Catalog No. 19-170-100 Supplier KleenGuard™ Supplier No. 25672
Add to Cart
Edge
Add to Cart
The KleenGuard Calico Safety Eyewear gives you and your workers a lightweight protective solution that offers excellent wrap around and anti-fog protection in a stylish and comfortable design.
  • Lightweight “wraparound” design for added comfort and protection
  • Impact resistant polycarbonate lens
  • Interchangeable head strap allows tight fit to user«s face
  • Interchangeable temples can be removed which allows product to be modified to the job
  • EN166 N anti-fog coating provides assured protection from fogging lens
  • Fog shield provides extra protection around lens
  • Cushioned brow bar for additional comfort and ventilation
  • Provides 99.9% UV protection, reduces eye strain and protects against radiation
Metal manufacturing, transportation, construction
Compliance
Meets EN166 1FT, CSA and ANSI™ Standards
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN166 1FT, CSA and ANSI Standards
Lens Coating Anti-fog
Lens Tint Clear
Product Type Safety Eyewear
Material Polycarbonate
UV Protection UV400