Learn More
Ansell KleenGuard™ V50 Epic™ Safety Eyewear
Catalog No. 19006210 Shop All Ansell Products
Change view
Click to view available options
Lens Tint:
Clear
Indoor/Outdoor
Smoke
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-006-210 Smoke
19-006-208 Clear
19-006-209 Indoor/Outdoor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-006-210 Supplier Ansell Supplier No. 35327
This item is not returnable. View return policy
KleenGuard™ V50 Epic™ Safety Eyewear feature a flexible foam gasket, which provides long term comfort and helps to seal out dust and debris.
  • Anti-fogging solution resists fogging up to two times longer than the EN166 industry standard
  • Direct ventilation at the top and bottom of the eyewear to provide superior anti-fog performance for worker efficiency and safety
  • Meets ANSI Z87.1+ standards and CSA Z94.3 standards
  • Lenses are ideal for indoor environments, thanks to their maximum light transmission
  • Temple bars can be interchanged with an included head strap when a closer, more secure fit is required
  • Wrap around design and impact resistant lenses make these ideal for use in dirty/metal manufacturing, transportation, as motorcycle goggles. etc.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Tint Smoke
Product Type Safety Eyewear
Frame Color Black
Temple Color Red
Lens Material Polycarbonate