Learn More
Ansell KleenGuard™ V50 OTG™ Safety Glasses
Catalog No. 19818754 Shop All Ansell Products
Change view
Click to view available options
Lens Tint:
Clear
Indoor/Outdoor, Indoor/Outdoor Mirror
Smoke
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-818754 Clear
19-818-755 Smoke
19-818-756 Indoor/Outdoor, Indoor/Outdoor Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-818754 Supplier Ansell Supplier No. 20746
Add to Cart
Add to Cart
This item is not returnable. View return policy
When you need to wear your prescription glasses and safety eye protection, you can trust KleenGuard OTG Over the Glasses Safety Glasses. They are made with a nylon frame for strength, durability and comfort and fit over most prescription glasses.
  • Made with a nylon frame for strength, durability and comfort and fit over most prescription glasses
  • See-through integrated side shields provide side protection without hindering peripheral vision
  • Hard coated for scratch resistance and longer life
  • Anti-fog coating for humid environment and temperature variances
  • Meet ANSI Z87.1+ standards for high impact
  • Polycarbonate lenses provide 99.9% UVA / UVB / UVC protection
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1 and CSA Z94.3
Lens Tint Clear
Product Type OTG safety spectacles
Frame Color Black
Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400