Description
- Ideal for when workers need vision assistance and eye protection
- Stylish, nylon frames feature a soft, self-adjusting rubber nose pad for comfort
- Clear lenses allow maximum visible light transmission (ideal for indoor use) and are hard coated for scratch resistance and longer life
- Meet ANSI Z87.1+ standards for high impact
- Polycarbonate lenses provide 99.9% UVA / UVB / UVC protection
- Come in three different strengths (+1.5, +2.0 and +2.5)
- 7 base curve, which helps provide for peripheral vision and protection and feature an hard coating to prevent lens damage
Specifications
Specifications
| Certifications/Compliance | ANSI Z87.1 |
| Lens Tint | Clear |
| Product Type | Dual Readers Safety Eyewear |
| For Use With (Application) | For performing tasks that require up and down near vision or working upside down or overhead |
| Frame Color | Black |
| Magnification Power | +2.00 |
| UV Protection | UV400 |