Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KC200 Isolation Cover Gowns

Catalog No. 18022870 Shop All Ansell Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-022-870 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-022-870 Supplier Ansell Supplier No. 69981

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Light fluid contact and contact isolation

Protective three-layer SMS fabric resistant to blood strike-through for various levels of exposure.

  • Elastic cuffs
  • Tape-tab neck closure
  • Ties at waist

OSHA requires the employer to evaluate the task and type of exposure anticipated and select the appropriate clothing that prevents contamination of an employee's skin or clothing by blood or other potentially infectious materials.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Healthcare, Medical, Clainical
Gender Unisex
No. of Pockets None
Size Universal
Product Type General Purpose Gown
Material Polypropylene
Color Blue
Closure Type Tape-tab at Neck
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Model 69981
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.