Learn More
Ansell KleenGuard™ A40 Shirts
Catalog No. 191202408B Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-2408B Medium
19-120-2408E 2X-Large
19-120-2408C Large
19-120-2408D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-120-2408B Supplier Ansell Supplier No. 44402
This item is not returnable. View return policy
Microporous film laminate XP liquid and particle protection shirts
  • Five snap front closure
  • Strong liquid and particulate barrier
  • Low in lint
  • Breathable
  • White
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Material Fabric
Color White
Closure Type Snaps
Certifications/Compliance ANSI ISEA 101-1996, NFPA 99, ASTM F1670
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.