Description
- Impact-resistant lenses offer 99.9% UVA/UVB protection
- Amber: For low-light environments where contrast is low; blocks out blue light to highlight and define shapes
- Smoke: For applications where eyestrain is a concern or bright, sunny days requiring maximum glare reduction
- Indoor/Outdoor: For moving between indoor/shaded areas to direct sunlight and back
V10 Standard Model
- Lightweight frames for added comfort
Compliance
Meet ANSI™ Z87.1-2003 standards.
Specifications
Specifications
| Lens Coating | Uncoated |
| Lens Tint | Amber |
| UV Protection | 0.999 |
| Frame Type | Lightweight |