Description
Made from a lightweight, nonwoven fabric laminated on the exterior with polyethylene film, with a soft, cloth-like feel on the interior for long-wearing comfort.
- Feature fuller cut in key stress areas for greater range of motion and easy mobility
- Protective enhancements include bound seams and the frontal primary splash area has no seams
- Ideal for environmental cleanup, acid handling, and biohazard cleanup
- Gray
Order Info
Choose from a variety of coverall styles. Apron and sleeve covers are also available.
Compliance
Meets ASTM F 903 penetration test standards.
Specifications
Specifications
| Size | Medium |
| Material | Composite fabric |
| Color | Gray |