Learn More
Ansell KleenGuard™ Light Duty Sleeve Protectors
GREENER_CHOICE
Catalog No. 19169971 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Large
Catalog No. Size
19-169-971 X-Large
1 options
This item is not returnable. View return policy
Catalog No. 19-169-971 Supplier Ansell Supplier No. 23610
Add to Cart
Add to Cart
This item is not returnable. View return policy
KleenGuard™ Light Duty Sleeve Protectors provide economical, disposable protection for workers who exposed to dirt and grime in the workplace
  • Designed for entry-level protection from hazardous dust
  • Lightweight polypropylene material is breathable for comfortable wear
  • Elastic on both the wrist and arm allows for a good fit
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Disposable Single-use
For Use With (Application) Fiberglass manufacturing, tank cleaning, utilities, liquid handling, pressure washing, maintenance, general manufacturing
Length (Metric) 45.7 cm