Learn More
Ansell KleenGuard™ Pre-Moistened Lens Cleaning Towelettes
Catalog No. 19413150 Shop All Ansell Products
Change view
Click to view available options
Quantity:
100/Box, 1000/Cs.
Catalog No. Quantity
19-413-150 100/Box, 1000/Cs.
1 options
Catalog No. 19-413-150 Supplier Ansell Supplier No. 14551
  • $451.00
    $343.65 / Case of 1000
    Save $107.35
    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Lens cleaning wipers pre-saturated with an anti-fog, anti-static formula. Comes in a wall-mountable corrugate box designed for easy wiper dispensing. There are 100 towelettes/box and 10 boxes/case.
  • 1,000 Pre-Moistened Lens Cleaning Towelettes / Case; 10 Boxes of 100 Towels
  • Lens cleaning wipers pre-saturated with an anti-fog, anti-static formula; Does not contain silicone
  • Comes in a wall-mountable corrugate box designed for easy wiper dispensing
  • Each towelette is individually packaged for cleanliness and convenience
  • Note: IPA-based lens cleaning towelettes can degrade eyewear anti-fog coating over time
TRUSTED_SUSTAINABILITY
Specifications
Product Type Lens Cleaning Towelette
Quantity 100/Box, 1000/Cs.