Learn More
KleenGuard™ Fluidshield™ Procedure Masks
Catalog No. 189994799 Shop All Ansell Products
Change view
Click to view available options
Fastening Type:
Earloops
Catalog No. Fastening Type
18-999-4799 Earloops
1 options
This item is not returnable. View return policy
Catalog No. 18-999-4799 Supplier KleenGuard™ Supplier No. 47117
This item is not returnable. View return policy
Face masks are constructed of fluid resistant materials and are lighter and cooler than average procedure masks.
  • Three-layer, pleat-style mask
  • Ear loops
  • Inner facing is light and breathable to reduce irritation
  • Outer facing resists fluid strike through
TRUSTED_SUSTAINABILITY
Specifications
Product Type Medical/Procedural Face Mask
Fastening Type Earloops
Sterility Non-sterile
Style Pleated
Filter Efficiency ≥97% (PFE), ≥96% (BFE)
Latex Free Yes
Resistance 160 mm Hg
Series Fluidshield™